Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- mthl2
- Entrez Gene ID
-
38636
- Gene Name
- methuselah-like 2
- Gene Aliases
-
CG17795, Dmel\CG17795, Dmel_CG17795, Mth-2, Mthl2, anon-WO0170980.28, anon-WO0170980.29
- Chromosome Location
-
Chr.3L: 5604790 - 5607190 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0035623
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_176284.3 |
NP_788462.2 |
6 - 7 |
1564 |
72 |
GenBank mRNA |
BT044127.1 |
- |
6 - 7 |
1565 |
72 |
View Full Product Details