Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bre1
Entrez Gene ID
38652
Gene Name
Bre1-PA
Gene Aliases
BRE1, Bre-1, CG10542, Dmel\CG10542, Dmel_CG10542, EC3-8, bre-1, bre1, dBRE1, dBre1, l(3)01640
Chromosome Location
Chr.3L: 5789831 - 5794192 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0086694
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300021.1 NP_001286950.1 3 - 4 2181 82
NM_139732.3 NP_647989.2 3 - 4 2181 82
GenBank mRNA BT082047.1 - 3 - 4 2180 82

View Full Product Details