Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Jon65Aiii
- Entrez Gene ID
-
38683
- Gene Name
- Jonah 65Aiii
- Gene Aliases
-
CG6483, Dmel\CG6483, Dmel_CG6483, Jon65, Jon65A, SP98, anon-EST:GressD16, anon-WO0140519.59, jon65Aiii
- Chromosome Location
-
Chr.3L: 6050757 - 6051690 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0035665
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_139756.3 |
NP_648013.1 |
1 - 2 |
282 |
64 |
GenBank mRNA |
AY095514.1 |
- |
1 - 2 |
265 |
64 |
View Full Product Details