Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Cpr65Az
- Entrez Gene ID
-
38712
- Gene Name
- Cuticular protein 65Az
- Gene Aliases
-
CG12330, DmelCpr65Az, Dmel\CG12330, Dmel_CG12330
- Chromosome Location
-
Chr.3L: 6160814 - 6161589 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0035686
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_139774.1 |
NP_648031.1 |
1 - 2 |
392 |
66 |
View Full Product Details