Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mthl7
Entrez Gene ID
38910
Gene Name
methuselah-like 7
Gene Aliases
CG7476, Dmel\CG7476, Dmel_CG7476, Mthl7, anon-WO0170980.85, anon-WO0170980.86
Chromosome Location
Chr.3L: 8178675 - 8180514 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0035847
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139924.3 NP_648181.2 2 - 3 845 107
NM_176297.1 NP_788475.1 3 - 4 797 107

View Full Product Details