Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ab
Entrez Gene ID
34560
Gene Name
abrupt
Gene Aliases
Ab, B2149, CG32830, CG43860, CG4807, Dmel\CG43860, Dmel_CG43860, clu, l(2)k02807, ptd
Chromosome Location
Chr.2L: 11210678 - 11261080 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0264442
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169481.2 NP_001162952.1 8 - 9 3017 98
NM_001273450.1 NP_001260379.1 6 - 7 2667 98
NM_057214.5 NP_476562.1 8 - 9 3069 98
NM_057215.5 NP_476563.1 8 - 9 3065 98

View Full Product Details