Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Acp26Ab
Entrez Gene ID
33816
Gene Name
Accessory gland protein 26Ab
Gene Aliases
Acp, Acp26A, Acp26Aab, Acp26a, CG9024, Dmel\CG9024, Dmel_CG9024, Mst26A, Mst26Ab, acp26Ab, msP355b, mst 355b, mst355b
Chromosome Location
Chr.2L: 5892298 - 5892862 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0002856
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298717.1 NP_001285646.1 1 - 2 58 58
NM_057297.3 NP_476645.1 1 - 2 58 58
NM_134272.1 NP_599099.1 1 - 2 58 58
GenBank mRNA BT023248.1 - 1 - 2 38 58
KX531361.1 - 1 - 2 33 58

View Full Product Details