Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RanGAP
Entrez Gene ID
35223
Gene Name
Ran GTPase activating protein
Gene Aliases
CG9999, CT28175, Dmel\CG9999, Dmel_CG9999, RGP1_DROME, Ran, RanGap, RanGap1, Rangap, SD, Scl, Sd, Sd-RanGAP, Sd-RanGap, dRanGAP, ranGAP, ranGap, rangap
Chromosome Location
Chr.2L: 19441959 - 19447317 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003346
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273649.1 NP_001260578.1 1 - 2 232 60
NM_001273650.1 NP_001260579.1 2 - 3 368 60
NM_057364.4 NP_476712.1 2 - 3 625 60
GenBank mRNA AY061219.1 - 1 - 2 217 60

View Full Product Details