Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
so
Entrez Gene ID
35662
Gene Name
sine oculis
Gene Aliases
CG11121, Dmel\CG11121, Dmel_CG11121, Drl, SO, Sine oculis, So, ami, mda, med, old, somda
Chromosome Location
Chr.2R: 7419350 - 7434872 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003460
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057385.5 NP_476733.1 6 - 7 1637 57
GenBank mRNA BT029256.1 - 6 - 7 1621 57
L31626.1 - 6 - 7 1953 57

View Full Product Details