Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sog
Entrez Gene ID
32498
Gene Name
short gastrulation
Gene Aliases
CG9224, Dm sog, Dmel\CG9224, Dmel_CG9224, SOG, Sog, l(1)G0160, l(1)G0395, l(1)G0479
Chromosome Location
Chr.X: 15603283 - 15633434 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003463
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272647.2 NP_001259576.1 2 - 3 1430 66
NM_001272649.2 NP_001259578.1 1 - 2 1112 66
NM_057388.4 NP_476736.1 1 - 2 1112 66
GenBank mRNA BT053679.1 - 2 - 3 1430 66
U18774.1 - 1 - 2 1457 66

View Full Product Details