Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SERCA
- Entrez Gene ID
-
49297
- Gene Name
- Sarco/endoplasmic reticulum Ca(2+)-ATPase
- Gene Aliases
-
17136664, A4UZU0_DROME, ATC1_DROME, CA-P60A, CG3725, Ca-ATPase, Ca-P60, Ca-P60A, Ca-p, CaP60A, Ca[2+]ATPase, Dmel\CG3725, Dmel_CG3725, Kum, SERCA2, Serca, Serca2a, dSERCA, dserca, kum, l(2)SH0211, l(2)k08308ab, l(2)k09025, serca
- Chromosome Location
-
Chr.2R: 23925077 - 23935884 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0263006
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057484.3 |
NP_476832.1 |
8 - 9 |
3647 |
141 |
View Full Product Details