The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_057485.4 | NP_476833.1 | 2 - 3 | 929 | 69 |
GenBank mRNA | AF132173.1 | - | 2 - 3 | 898 | 69 |
BT010045.1 | - | 2 - 3 | 901 | 69 |