Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TfIIB
Entrez Gene ID
34430
Gene Name
Transcription factor IIB
Gene Aliases
CG5193, Dmel\CG5193, Dmel_CG5193, TFIIB, Tfiib, TfllB, dTFIIB, dTfIIB, dmTFIIB, tfIIb
Chromosome Location
Chr.2L: 10431338 - 10433116 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0004915
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273420.1 NP_001260349.1 2 - 3 490 62
NM_057540.4 NP_476888.1 2 - 3 490 62
GenBank mRNA BT011459.1 - 2 - 3 484 62
BT015269.1 - 2 - 3 452 62
M88164.1 - 2 - 3 473 62
M91081.1 - 2 - 3 469 62

View Full Product Details