Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
capu
Entrez Gene ID
33611
Gene Name
cappuccino
Gene Aliases
CG15420, CG3399, Capu, Dmel\CG3399, Dmel_CG3399, capp
Chromosome Location
Chr.2L: 3872646 - 3902860 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0000256
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144311.2 NP_001137783.1 7 - 8 3699 68
NM_001144312.2 NP_001137784.1 7 - 8 3942 68
NM_001144313.2 NP_001137785.1 4 - 5 2450 68
NM_001144314.2 NP_001137786.1 5 - 6 3118 68
NM_001144315.2 NP_001137787.1 6 - 7 3691 68
NM_057618.3 NP_476966.1 6 - 7 3266 68
NM_164565.2 NP_722950.2 6 - 7 3126 68
NM_164566.3 NP_722951.1 7 - 8 3710 68
NM_164567.2 NP_722952.1 6 - 7 3034 68
GenBank mRNA AY119486.1 - 6 - 7 3034 68
BT099743.1 - 4 - 5 2421 68

View Full Product Details