Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- a5
- Entrez Gene ID
-
33317
- Gene Name
- antennal protein 5
- Gene Aliases
-
A5, CG5430, Dmel\CG5430, Dmel_CG5430
- Chromosome Location
-
Chr.2L: 1329161 - 1329993 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0011294
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057650.3 |
NP_476998.1 |
1 - 2 |
160 |
83 |
View Full Product Details