The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001273063.1 | NP_001259992.1 | 2 - 3 | 803 | 91 |
NM_057669.3 | NP_477017.1 | 2 - 3 | 628 | 91 | |
GenBank mRNA | BT004845.1 | - | 2 - 3 | 627 | 91 |
U10328.1 | - | 2 - 3 | 618 | 91 |