Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
igl
Entrez Gene ID
36681
Gene Name
igloo
Gene Aliases
CG18285, Dmel\CG18285, Dmel_CG18285, Igl
Chromosome Location
Chr.2R: 15155437 - 15190292 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0013467
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057712.5 NP_477060.1 1 - 2 781 71
GenBank mRNA S72573.1 - 1 - 2 720 71

View Full Product Details