Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
drl
Entrez Gene ID
44355
Gene Name
derailed
Gene Aliases
CG10758, CG17348, DRL, Dmel\CG17348, Dmel_CG17348, Drl, lin, lio, lio/drl
Chromosome Location
Chr.2L: 19190342 - 19208621 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0015380
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273637.1 NP_001260566.1 3 - 4 2103 66
NM_057791.4 NP_477139.1 3 - 4 2103 66
GenBank mRNA AY051852.1 - 3 - 4 2084 66
L47260.1 - 3 - 4 2064 66
U36584.1 - - 2094 66

View Full Product Details