Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
toc
Entrez Gene ID
33526
Gene Name
toucan
Gene Aliases
BA34, BcDNA:LD27161, CG9660, Dmel\CG9660, Dmel_CG9660, Toc, l(2)01361, l(2)05527, l(2)K08224, l(2)k08224
Chromosome Location
Chr.2L: 3068345 - 3144786 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0015600
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057805.3 NP_477153.1 4 - 5 4217 75
NM_164524.2 NP_722875.2 2 - 3 381 75
GenBank mRNA BT031283.1 - 2 - 3 365 75

View Full Product Details