Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- T3dh
- Entrez Gene ID
-
37551
- Gene Name
- Type III alcohol dehydrogenase
- Gene Aliases
-
CG3425, Dmel\CG3425, Dmel_CG3425
- Chromosome Location
-
Chr.2R: 22329726 - 22332072 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0017482
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057861.3 |
NP_477209.2 |
4 - 5 |
677 |
63 |
View Full Product Details