Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SA
- Entrez Gene ID
-
33974
- Gene Name
- Stromalin
- Gene Aliases
-
CG3423, DSA, DSA1, DmSA, Dmel\CG3423, Dmel_CG3423, ESTS:92H2T, SA-1, SCC3, Scc3, stromalin
- Chromosome Location
-
Chr.2L: 6950421 - 6954586 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0020616
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057920.4 |
NP_477268.2 |
4 - 5 |
982 |
102 |
GenBank mRNA |
AY061410.1 |
- |
4 - 5 |
963 |
102 |
View Full Product Details