Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rack1
Entrez Gene ID
34070
Gene Name
Receptor of activated protein kinase C 1
Gene Aliases
CG7111, Dmel\CG7111, Dmel_CG7111, RACK1, dRack1, i173, rack1
Chromosome Location
Chr.2L: 7825347 - 7827315 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0020618
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273289.1 NP_001260218.1 2 - 3 402 65
NM_001298794.1 NP_001285723.1 2 - 3 471 65
NM_057921.4 NP_477269.1 2 - 3 402 65
GenBank mRNA AY071661.1 - 2 - 3 435 65

View Full Product Details