Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Orc1
- Entrez Gene ID
-
35686
- Gene Name
- Origin recognition complex subunit 1
- Gene Aliases
-
CG10667, DmORC1, DmOrc1, DmOrc1p, Dmel\CG10667, Dmel_CG10667, ORC, ORC1, dORC1, fs(2)SC46, orc1, rDmORC, sat
- Chromosome Location
-
Chr.2R: 7522437 - 7525716 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0286788
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057955.4 |
NP_477303.1 |
2 - 3 |
2317 |
78 |
GenBank mRNA |
AY094780.1 |
- |
2 - 3 |
2298 |
78 |
View Full Product Details