Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scw
Entrez Gene ID
46000
Gene Name
screw
Gene Aliases
BcDNA:LD30945, CG31695, Dmel\CG31695, Dmel_CG31695, SCW, Scw, l(2)37Ff, l(2)IG76, mor2
Chromosome Location
Chr.2L: 19699379 - 19700968 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0005590
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299159.1 NP_001286088.1 1 - 1 1236 94
NM_080124.5 NP_524863.3 1 - 1 1236 94
GenBank mRNA AY051793.1 - 1 - 1 1200 94

View Full Product Details