Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Klp54D
Entrez Gene ID
47216
Gene Name
Kinesin-like protein at 54D
Gene Aliases
CG15844, CG43323, DmKlp54D, Dmel\CG43323, Dmel_CG43323, KIF 12, KIF12, KLP1, KLP54D, Klp1
Chromosome Location
Chr.2R: 17557031 - 17563406 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0263076
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001316425.1 NP_001303354.1 6 - 7 2038 74
NM_001316426.1 NP_001303355.1 6 - 7 2038 74
NM_080144.3 NP_524883.2 6 - 7 2038 74

View Full Product Details