Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tsp3A
Entrez Gene ID
31244
Gene Name
Tetraspanin 3A
Gene Aliases
BACR7C10.6, CG10742, Dm.Tsp3A, Dmel\CG10742, Dmel_CG10742, EG:BACR7C10.6, tsp3A
Chromosome Location
Chr.X: 2617758 - 2619738 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0040334
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258575.1 NP_001245504.1 2 - 3 789 64
NM_080315.3 NP_525054.2 2 - 3 789 64
GenBank mRNA BT060443.1 - 2 - 3 800 64

View Full Product Details