Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Crag
Entrez Gene ID
31800
Gene Name
Calmodulin-binding protein related to a Rab3 GDP/GTP exchange protein
Gene Aliases
CG12737, CJ, CRAG, Dmel\CG12737, Dmel_CG12737
Chromosome Location
Chr.X: 8592031 - 8602036 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0025864
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169239.2 NP_001162710.2 10 - 11 4198 107
NM_080341.4 NP_525080.3 10 - 11 4198 107

View Full Product Details