Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Samuel
Entrez Gene ID
34530
Gene Name
SAM-motif ubiquitously expressed punctatedly localized protein
Gene Aliases
78BP, CG12291, CG14917, CG14918, CG31868, Dmel\CG31868, Dmel_CG31868, Moses, mdcds_1002, moses
Chromosome Location
Chr.2L: 11013206 - 11066998 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0032330
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273443.3 NP_001260372.1 6 - 7 3607 68
NM_080362.4 NP_525101.2 6 - 7 3008 68

View Full Product Details