Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
upd3
Entrez Gene ID
3346149
Gene Name
unpaired 3
Gene Aliases
CG15062, CG15062/CG5963, CG33542, CG5963, Dmel\CG33542, Dmel_CG33542, UPD3, Upd, Upd-3, Upd-like, Upd3, cg15062, cg5963, od, odsy, os, sy, upd, upd 3, upd-3
Chromosome Location
Chr.X: 18277233 - 18284596 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0053542
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103544.2 NP_001097014.1 2 - 3 819 58
GenBank mRNA BT044177.1 - 2 - 3 819 58

View Full Product Details