Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- eIF6
- Entrez Gene ID
-
37776
- Gene Name
- eukaryotic translation initiation factor 6
- Gene Aliases
-
CG17611, DeIF6, Dmel\CG17611, Dmel_CG17611, anon-WO0172774.153, anon-WO0172774.155, eif6, elF6, l(2)k13214
- Chromosome Location
-
Chr.2R: 23849246 - 23850301 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0034915
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_145105.3 |
NP_659573.1 |
1 - 1 |
943 |
65 |
GenBank mRNA |
AY094688.1 |
- |
1 - 1 |
866 |
65 |
View Full Product Details