Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NLaz
Entrez Gene ID
33324
Gene Name
Neural Lazarillo
Gene Aliases
BcDNA:RE67583, CG16933, CG33126, CT33980, DNLaz, Dmel\CG33126, Dmel_CG33126, Nlaz
Chromosome Location
Chr.2L: 1359837 - 1361821 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0053126
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272938.2 NP_001259867.1 2 - 3 220 63
NM_001298631.1 NP_001285560.1 2 - 3 306 63
NM_175946.2 NP_787960.1 2 - 3 220 63
GenBank mRNA AY084185.1 - 2 - 3 217 63

View Full Product Details