Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
b6
Entrez Gene ID
31124
Gene Name
CG3100-PA
Gene Aliases
B6, CG3100, Dmel\CG3100, Dmel_CG3100, EG:9D2.1
Chromosome Location
Chr.X: 1686624 - 1690187 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0024897
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058017.5 NP_477365.1 4 - 5 1135 93
GenBank mRNA BT050466.1 - 4 - 5 1133 93

View Full Product Details