Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NTPase
Entrez Gene ID
33495
Gene Name
CD39-like NTPase
Gene Aliases
CG3059, Dmel\CG3059, Dmel_CG3059, NTPDase6, dmNTPase
Chromosome Location
Chr.2L: 2880704 - 2885850 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0024947
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273043.1 NP_001259972.1 2 - 3 1674 77
NM_058022.5 NP_477370.1 3 - 4 1479 77
NM_164513.4 NP_722860.1 3 - 4 1494 77
NM_164514.3 NP_722861.1 2 - 3 1565 77
NM_164515.3 NP_722862.1 3 - 4 1490 77
GenBank mRNA AY061134.1 - 3 - 4 1466 77
BT133416.1 - 3 - 4 1488 77
EU852293.1 - 3 - 4 1293 77

View Full Product Details