The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_058026.4 | NP_477374.1 | 2 - 3 | 1987 | 72 |
GenBank mRNA | AJ010169.1 | - | 2 - 3 | 1981 | 72 |