Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PpN58A
Entrez Gene ID
37493
Gene Name
Protein phosphatase N at 58A
Gene Aliases
CG3245, DmPpN-58A, Dmel\CG3245, Dmel_CG3245, PPN, PPN 58A, PpN-58A
Chromosome Location
Chr.2R: 21881352 - 21882533 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0025573
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058036.4 NP_477384.1 1 - 1 1075 132
GenBank mRNA BT083436.1 - 1 - 1 1075 132

View Full Product Details