Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pten
Entrez Gene ID
43991
Gene Name
Phosphatase and tensin homolog
Gene Aliases
CG5671, D.PTEN, DPTEN, Dmel\CG5671, Dmel_CG5671, PTEN, PTEN3, dPTEN, dPten, dpten, pten
Chromosome Location
Chr.2L: 10256319 - 10261049 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0026379
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169462.2 NP_001162933.1 7 - 8 1786 155
NM_058074.4 NP_477422.2 8 - 9 1629 155
NM_058075.4 NP_477423.1 8 - 9 1517 155
NM_058076.4 NP_477424.1 6 - 7 1287 155
NM_134320.3 NP_599147.1 8 - 9 1517 155
NM_134321.3 NP_599148.1 6 - 7 1287 155
NM_134322.3 NP_599149.1 6 - 7 1287 155
GenBank mRNA AF144232.1 - 7 - 8 1060 155
AF161257.1 - 6 - 7 1298 155
AF161259.1 - 6 - 7 1298 155
BT025053.1 - 7 - 8 1715 155

View Full Product Details