Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- amos
- Entrez Gene ID
-
35110
- Gene Name
- absent MD neurons and olfactory sensilla
- Gene Aliases
-
Amos, CG10393, Dmel\CG10393, Dmel_CG10393, Roi, Tft, asolo, bHLHa10, rolo
- Chromosome Location
-
Chr.2L: 18596136 - 18596921 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0003270
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_058098.3 |
NP_477446.1 |
1 - 1 |
664 |
81 |
View Full Product Details