Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dnz1
Entrez Gene ID
34503
Gene Name
DNZDHHC/NEW1 zinc finger protein 11
Gene Aliases
CG6627, DNZ1, Dmel\CG6627, Dmel_CG6627
Chromosome Location
Chr.2L: 10854435 - 10856644 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0027453
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058101.4 NP_477449.1 1 - 2 486 66
GenBank mRNA AY061178.1 - 1 - 2 475 66

View Full Product Details