Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ft
- Entrez Gene ID
-
33627
- Gene Name
- fat
- Gene Aliases
-
79/18, CG3352, CT11259, Dm Fat, Dmel\CG3352, Dmel_CG3352, FT, Fat, Ft, l(2)24Da, l(2)79/18, l(2)fat, l(2)fd, l(2)ft, l(2)gd-1, l(2)gd-l, l(2)gd2, l(2)k07918
- Chromosome Location
-
Chr.2L: 4198404 - 4221796 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0001075
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_058149.4 |
NP_477497.1 |
3 - 4 |
6418 |
79 |
View Full Product Details