Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MKP-4
Entrez Gene ID
32963
Gene Name
MAPK Phosphatase 4
Gene Aliases
CG14211, DMKP-4, Dmel\CG14211, Dmel_CG14211, MAPK, Mkp4
Chromosome Location
Chr.X: 19623114 - 19625729 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0031044
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272788.1 NP_001259717.1 1 - 2 231 69
NM_134488.2 NP_608332.2 1 - 2 244 69
GenBank mRNA BT031134.1 - 1 - 2 208 69

View Full Product Details