Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RunxB
Entrez Gene ID
33051
Gene Name
Runt related B
Gene Aliases
CG15454, CG15455, CG42267, Dmel\CG42267, Dmel_CG42267
Chromosome Location
Chr.X: 20538497 - 20567097 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0259162
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272816.2 NP_001259745.1 5 - 6 1250 126

View Full Product Details