Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- mthl13
- Entrez Gene ID
-
246395
- Gene Name
- methuselah-like 13
- Gene Aliases
-
CG30018, Dmel\CG30018, Dmel_CG30018, Mthl13
- Chromosome Location
-
Chr.2R: 10791459 - 10793109 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0050018
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_176145.3 |
NP_788325.2 |
2 - 3 |
513 |
127 |
View Full Product Details