Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
inaE
Entrez Gene ID
32343
Gene Name
inactivation no afterpotential E
Gene Aliases
CG11096, CG11102, CG11104, CG15761, CG32630, CG33174, DAGL, Dmel\CG33174, Dmel_CG33174, EP(X)1101
Chromosome Location
Chr.X: 13783714 - 13811493 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0261244
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144723.2 NP_001138195.1 13 - 14 2737 70
NM_001144724.2 NP_001138196.1 13 - 14 2737 70
NM_176727.3 NP_788900.1 13 - 14 2650 70
GenBank mRNA EU817580.1 - 13 - 14 2615 70

View Full Product Details