Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
inaE
Entrez Gene ID
32343
Gene Name
inactivation no afterpotential E
Gene Aliases
CG11096, CG11102, CG11104, CG15761, CG32630, CG33174, DAGL, Dmel\CG33174, Dmel_CG33174, EP(X)1101
Chromosome Location
Chr.X: 13783714 - 13811493 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0261244
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144723.2 NP_001138195.1 6 - 7 1211 66
NM_001144724.2 NP_001138196.1 6 - 7 1211 66
NM_001298301.1 NP_001285230.1 6 - 7 1211 66
NM_001298302.1 NP_001285231.1 6 - 7 1211 66
NM_176727.3 NP_788900.1 6 - 7 1124 66
NM_176728.3 NP_788901.1 6 - 7 1211 66
GenBank mRNA AY122194.1 - 6 - 7 1089 66
EU817579.1 - 6 - 7 1089 66
EU817580.1 - 6 - 7 1089 66

View Full Product Details