Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- AnxB9
- Entrez Gene ID
-
42492
- Gene Name
- Annexin B9
- Gene Aliases
-
ANXB9, Ann, AnnB9, AnnIX, Anx B9, CG5730, Dmel\CG5730, Dmel_CG5730, annexin
- Chromosome Location
-
Chr.3R: 21065241 - 21070917 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0000083
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057256.4 |
NP_476604.1 |
4 - 5 |
994 |
76 |
View Full Product Details