Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tok
Entrez Gene ID
42944
Gene Name
tolkin
Gene Aliases
CG6863, CT21251, Dmel\CG6863, Dmel_CG6863, TLR, TOK, Tlr, Tlr-1, Tok, tldr-1, tldr1, tlr, tlr-1, tlr1
Chromosome Location
Chr.3R: 24719697 - 24749125 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0004885
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300581.1 NP_001287510.1 4 - 5 1553 56
NM_057531.4 NP_476879.1 4 - 5 1557 56
NM_170168.2 NP_733035.1 4 - 5 1436 56
GenBank mRNA AY051585.1 - 4 - 5 1543 56

View Full Product Details