Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ppan
- Entrez Gene ID
-
42502
- Gene Name
- peter pan
- Gene Aliases
-
CG5786, Dmel\CG5786, Dmel_CG5786, PPAN, Ppan, l(3)02231, l(3)j6B6
- Chromosome Location
-
Chr.3R: 21116254 - 21117899 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0010770
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057631.6 |
NP_476979.1 |
3 - 4 |
1168 |
72 |
View Full Product Details