Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tmod
Entrez Gene ID
43633
Gene Name
tropomodulin
Gene Aliases
1309/10, CG11493, CG1539, CG15540, CT3919, CT41421, Dmel\CG1539, Dmel_CG1539, E42, Tmod, l(3)00848, l(3)S130910, spdo
Chromosome Location
Chr.3R: 30532607 - 30578905 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0082582
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104512.4 NP_001097982.1 3 - 4 604 77
NM_001170304.3 NP_001163775.1 2 - 3 636 77
NM_001170305.3 NP_001163776.1 3 - 4 919 77
NM_001170306.2 NP_001163777.1 3 - 4 672 77
NM_001202390.1 NP_001189319.1 3 - 4 634 77
NM_001202391.2 NP_001189320.1 3 - 4 581 77
NM_001260442.2 NP_001247371.1 3 - 4 598 77
NM_001260443.2 NP_001247372.1 3 - 4 712 77
NM_001260444.2 NP_001247373.1 3 - 4 665 77
NM_001276182.1 NP_001263111.1 2 - 3 584 77
NM_001276183.1 NP_001263112.1 2 - 3 584 77
NM_001382033.1 NP_001369033.1 3 - 4 604 77
NM_057683.5 NP_477031.1 2 - 3 456 77
NM_057684.4 NP_477032.1 2 - 3 427 77
NM_170503.4 NP_733382.1 2 - 3 825 77
NM_170504.4 NP_733383.1 3 - 4 604 77
NM_170505.4 NP_733384.1 3 - 4 689 77
NM_170506.3 NP_733385.1 3 - 4 635 77
GenBank mRNA AY094727.1 - 2 - 3 427 77
BT099728.1 - 3 - 4 635 77
BT100046.1 - 3 - 4 673 77
BT133205.1 - 2 - 3 428 77
BT133473.1 - 3 - 4 597 77
U92490.1 - 2 - 3 465 77

View Full Product Details