Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Dfd
- Entrez Gene ID
-
40832
- Gene Name
- Deformed
- Gene Aliases
-
BG:DS00276.5, CG2189, DFD, DmDfd, Dmel\CG2189, Dmel_CG2189, EbR11, dfd, l(3)84Ae
- Chromosome Location
-
Chr.3R: 6791836 - 6802428 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0000439
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057853.3 |
NP_477201.1 |
4 - 5 |
1938 |
124 |
View Full Product Details