Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tacc
Entrez Gene ID
40589
Gene Name
transforming acidic coiled-coil protein
Gene Aliases
CG9765, D-TACC, D-tacc, DTACC, Dmel\CG9765, Dmel_CG9765, TACC, TACCdm, acc, d-tacc, dTACC, dtacc
Chromosome Location
Chr.3R: 4733654 - 4749056 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0026620
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202240.2 NP_001189169.1 1 - 2 619 60
NM_001275332.1 NP_001262261.1 1 - 2 619 60
NM_058085.5 NP_477433.2 1 - 2 619 60
GenBank mRNA AY058499.1 - 1 - 2 619 60

View Full Product Details